Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
5 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 304,00 zł |
Informacje o tej pozycji
304,00 zł
material
green polypropylene
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120202
well diam.
12.3 mm
Features and Benefits
- for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
- easy-open hinged lid with snap clasp
- autoclavable
- designed for quick visual identification
- uniform wells with location indentification
- tube capacity is indicated on the outside of the box
- translucent vibrant colors with frosted labeling area
- debossed with a distinctive wave design
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description green polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes | description orange polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes | description yellow polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes | description Holds 100 x 1.5-2.0 mL tubes, black polypropylene, pk of 5 |
| manufacturer/tradename Heathrow Scientific HS120202 | manufacturer/tradename Heathrow Scientific HS120204 | manufacturer/tradename Heathrow Scientific HS120203 | manufacturer/tradename Heathrow Scientific HS120043 |
| feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes | feature Holds 100 x 1.5-2.0 mL tubes |
| material green polypropylene | material orange polypropylene | material yellow polypropylene | material black polypropylene |
| packaging pk of 5 | packaging pk of 5 | packaging pk of 5 | packaging pk of 5 |
| well diam. 12.3 mm | well diam. 12.3 mm | well diam. 12.3 mm | well diam. 12.3 mm |
Still not finding the right product?
Explore all of our products under Microtube Storage Box
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?