Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 6330,00 zł |
Informacje o tej pozycji
6330,00 zł
feature
to hold 4 x 1,000 mL buckets, to hold 4 x plate/tube buckets, swing-bucket rotor, without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895101003 (Global)
parameter
3,153 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description swing-bucket rotor, S-4x1000, to hold, 4 x 1,000 mL buckets, without buckets | description swing-bucket rotor, S-4x1000, aerosol-tight, to hold, 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid | description swing-bucket rotor, S-4x1000, aerosol-tight, with 4 x universal-large buckets | description FA-48x2, aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, with Eppendorf QuickLock rotor lid |
| compatibility for use with centrifuge models 5920R | compatibility for use with centrifuge models 5920R | compatibility for use with centrifuge models 5910R | compatibility for use with centrifuge models 5920R |
| manufacturer/tradename Eppendorf® 5895101003 (Global) | manufacturer/tradename Eppendorf® 5895100007 (Global) | manufacturer/tradename Eppendorf® 5895190006 (Global) | manufacturer/tradename Eppendorf® 5895135005 (Global) |
| parameter 3,153 × g max. RCF | parameter 3,428 × g max. RCF | parameter 4,200 × g max. RCF | parameter 20,913 × g max. RCF |
| feature swing-bucket rotor, without buckets, to hold 4 x 1,000 mL buckets, to hold 4 x plate/tube buckets | feature to hold 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), with 4 × 1000 mL round buckets, aerosol-tight, swing-bucket rotor, with Eppendorf QuickLock® lid | feature aerosol-tight, with 4 x universal-large buckets, swing-bucket rotor, to hold 0.2 to 750 mL ubes and flasks and plates | feature with Eppendorf QuickLock rotor lid, aerosol tight, to hold 48 x 1.5/2.0 mL tubes, fixed angle rotor |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® 5920R Centrifuges
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?