Przejdź do
EP0030127790
Eppendorf® Microplate Sealing Films & Foils
PCR foil, PCR clean, autoclavable, self-adhesive, pkg of 100 ea
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
100 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
753,00 zł
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Eppendorf® 0030127790
Sterility:
non-sterile
Packaging:
pkg of 100 ea
753,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
sterility
non-sterile
feature
PCR clean, autoclavable, PCR foil, self-adhesive
packaging
pkg of 100 ea
manufacturer/tradename
Eppendorf® 0030127790
technique(s)
PCR: suitable
General description
To protect from evaporation and contamination, Eppendorf deepwell and microplates can be closed with different systems. Storage film and foil adhesive closurs close Eppendorf deepwell and microplates in an easy and reliable way. Film products are transparent and protect from accidental penetration. Foil products are made of aluminum and protect light sensitive samples.
Note: For long term storage the use of Eppendorf heat sealing film is recommended.
Application
Single use Eppendorf sealing films and foils are designed to seal deepwell and microplate to protect from evaporation and contamination.
Features and Benefits
  • Heat Sealing Film and Foil for continuous locking with the best evaporation protection
  • Easy removal without leaving behind any residue
  • Airtight self-adhesive films prevent sample loss due to evaporation
  • Highly chemical resistant to most of the substances in the laboratory
  • Foil products
    • Made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products
    • Transparent and provide protection against unwanted punctures
    • Transparent sealing film offers better visual clarity and helps in the monitoring of samples
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
EP0030127781EP0030127870BR701431
description
PCR foil, PCR clean, autoclavable, self-adhesive, pkg of 100 ea
description
PCR film, PCR clean, autoclavable, self-adhesive, pkg of 100 ea
description
self-adhesive, pkg of 100 ea, PCR clean
description
qPCR, suitable for PCR, storage, PE/PP/adhesive, operating temperature (-80)—110 °C, pack of 100 ea, transparent
sterility
non-sterile
sterility
non-sterile
sterility
-
sterility
non-sterile
manufacturer/tradename
Eppendorf® 0030127790
manufacturer/tradename
Eppendorf® 0030127781
manufacturer/tradename
Eppendorf® 0030127870
manufacturer/tradename
BRAND 701431
packaging
pkg of 100 ea
packaging
pkg of 100 ea
packaging
pkg of 100 ea
packaging
pack of 100 ea
technique(s)
PCR: suitable
technique(s)
PCR: suitable
technique(s)
-
technique(s)
-
feature
PCR foil, PCR clean
feature
PCR clean, self-adhesive, autoclavable, PCR film
feature
self-adhesive, PCR clean
feature
RNase and DNase free, qPCR, DNA free
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)