Przejdź do
NA12/22
Diaphragm for NOVASEPTIC® Valve
PTFE
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
31411800
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
EPDM rubber device (ethylene propylene diene monomer rubber), PTFE
feature
hydrophobic (Surface)
parameter
-30-140 °C temp. range (Short term use), -6-(-1) bar pressure range
weight
0.01 kg (0.022 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
NA12/25NA76/25NA18/25
material
EPDM rubber device (ethylene propylene diene monomer rubber), PTFE
material
PTFE , PTFE device (polytetrafluoroethylene), valve (Diaphragm, PTFE)
material
PTFE , PTFE device (polytetrafluoroethylene)
material
PTFE , PTFE device (polytetrafluoroethylene)
weight
0.01 kg (0.022 lb)
weight
0.01 kg (0.022 lb)
weight
0.16 kg (0.353 lb)
weight
0.02 kg (0.044 lb)
Quality Level
SP2
Quality Level
SP2
Quality Level
SP2
Quality Level
SP2
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
parameter
-30-140 °C temp. range (Short term use)
parameter
-40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use)
parameter
-40-170 °C temp. range (Continuous use), -40-250 °C temp. range (Short term use)
parameter
-40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use)
Still not finding the right product?
Explore all of our products under Diaphragm for NOVASEPTIC® Valve