Informacje o tej pozycji
UNSPSC Code:
31411800
Pomoc techniczna
Pozwól nam pomócQuality Segment
material
PTFE , PTFE device (polytetrafluoroethylene)
feature
hydrophobic (Surface)
parameter
-40-170 °C temp. range (Continuous use), -40-250 °C temp. range (Short term use)
weight
0.16 kg (0.353 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Diaphragm for NOVASEPTIC® Valve