Przejdź do
SYNF414400
Synthware™ two neck round bottom flask with angled side neck
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonim(y):
Synthware F414400, 2-neck round flask
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
100 mL, 100 mL
Material:
borosilicate glass flask, round bottom , two-neck (angled side neck)
Joint:
center joint: ST/NS24/40, side joint: ST/NS24/40
Packaging:
pkg of 1 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
borosilicate glass flask, round bottom , two-neck (angled side neck)
joint
center joint: ST/NS24/40, side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F414400
capacity
100 mL, 100 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
SYNF414100SYNF414504SYNF411100
description
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
description
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
description
500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
description
100 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
capacity
100 mL , 100 mL
capacity
100 mL , 100 mL
capacity
50 mL , 500 mL, 500 mL
capacity
100 mL , 100 mL
manufacturer/tradename
Synthware F414400
manufacturer/tradename
Synthware F414100
manufacturer/tradename
Synthware F414504
manufacturer/tradename
Synthware F411100
material
borosilicate glass flask, round bottom , two-neck (angled side neck)
material
two-neck (angled side neck), round bottom , borosilicate glass flask
material
two-neck (angled side neck), borosilicate glass flask, round bottom
material
two-neck , borosilicate glass flask, round bottom
joint
side joint: ST/NS24/40, center joint: ST/NS24/40
joint
side joint: ST/NS14/20, center joint: ST/NS24/40
joint
center joint: ST/NS24/40, side joint: ST/NS24/40
joint
center joint: ST/NS14/20, side joint: ST/NS14/20
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)