Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 285,00 zł |
Informacje o tej pozycji
285,00 zł
material
borosilicate glass flask, round bottom , two-neck (angled side neck)
joint
center joint: ST/NS24/40, side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F414100
capacity
100 mL, 100 mL
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 100 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20 | description 100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description 100 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 | description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20 |
| manufacturer/tradename Synthware F414100 | manufacturer/tradename Synthware F414400 | manufacturer/tradename Synthware F411100 | manufacturer/tradename Synthware F414150 |
| capacity 100 mL , 100 mL | capacity 100 mL , 100 mL | capacity 100 mL , 100 mL | capacity 50 mL , 50 mL |
| material two-neck (angled side neck), borosilicate glass flask, round bottom | material borosilicate glass flask, two-neck (angled side neck), round bottom | material two-neck , borosilicate glass flask, round bottom | material round bottom , two-neck (angled side neck), borosilicate glass flask |
| joint center joint: ST/NS24/40, side joint: ST/NS14/20 | joint side joint: ST/NS24/40, center joint: ST/NS24/40 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint side joint: ST/NS14/20, center joint: ST/NS24/40 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ two neck round bottom flask with angled side neck
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?