Yes, the glassy carbon electrode (GCE) is suitable for stability testing at negative potentials of -1 to -1.5 V, including applications such as chronopotentiometry. It can effectively serve as a counter electrode in hydrogen evolution reactions (HER) using 1 M KOH.
Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1220,00 zł |
Informacje o tej pozycji
1220,00 zł
product line
SynLectro®
size
3 mm × 20 mm × 60 mm
General description
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm |
| size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm |
| product line SynLectro® | product line SynLectro® | product line SynLectro® | product line SynLectro® |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
-
Is this electrode suitable for stability testing at negative potentials of -1 to-1.5 V, like chronopotentiometry testing? I want to use it as counter electrode in HER using 1 M KOH.
1 answer-
Helpful?
-
-
Is there any kind of documentation on the elemental and/or structural analysis of the electrode? I.e. what percentage is amorphous carbon vs graphite?
1 answer-
There is no documentation on the elemental and/or structural analysis of the electrode, including the percentages of amorphous carbon versus graphite. This product is made from glassy carbon, which is a non-graphitizing carbon material.
Helpful?
-
-
Is it possible to cut the electrode ESYNTH019-1EA into smaller pieces, for example, cutting it in half to make 3mm x 10mm x 30mm from the original 3 mm × 20 mm × 60 mm? If so, what is the best method for doing so - diamond scribe or some other method?
1 answer-
In the past, diamond scribers have been used for this purpose. However, this method requires quickly pressing an edge against it on the reverse side. If the customer has access to a mechanical workshop, a diamond saw is recommended. It should be noted that glassy carbon becomes harder with higher pressure. Therefore, when using a diamond saw, it is advisable to saw slowly and not apply excessive pressure.