Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 28,00 zł |
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41122403
Manufacturer/tradename:
BRAND 136100
Material:
high-density polyethylene
28,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
high-density polyethylene
description
with steel core
packaging
pack of 1 ea
manufacturer/tradename
BRAND 136100
L × D
260 mm × 7 mm
General description
The BRAND® stirring spatula features a PE-HD coated steel core, providing durability and strength for various laboratory applications. With two differently shaped ends, each approximately 16 mm wide, this spatula is designed for effective mixing, stirring, and scraping of both solid and liquid substances. The PE-HD coating ensures resistance to chemicals and prevents contamination during use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description high-density polyethylene | description polystyrene | description polypropylene | description PTFE (with spatulate ends), L × D 200 mm × 8 mm |
| manufacturer/tradename BRAND 136100 | manufacturer/tradename BRAND 759800 | manufacturer/tradename BRAND 136110 | manufacturer/tradename BRAND 136215 |
| material high-density polyethylene | material polystyrene | material polypropylene | material PTFE (with spatulate ends) |
| L × D 260 mm × 7 mm | L × D - | L × D 245 mm × 6 mm | L × D 200 mm × 8 mm |
| description with steel core | description - | description - | description with spatulate ends |
| packaging pack of 1 ea | packaging pack of 500 ea | packaging pack of 10 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under BRAND® stirring spatula
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?