Skip to Content
Merck
All Photos(1)

Key Documents

808027

TSKgel® GMPW (17 µm) HPLC Columns

L x I.D. 60 cm × 7.5 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
PLN 14,600.00

PLN 14,600.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
PLN 14,600.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

PLN 14,600.00


Please contact Customer Service for Availability

Product Name

TSKgel® GMPW HPLC Column, phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 17 μm particle size

material

stainless steel hardware

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 08027

parameter

≤0.5 M salt concentration
≤20% organic solvent
20 bar max. pressure
8 mL/min flow rate
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

60 cm × 7.5 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

17 μm

pore size

<100 Å

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel GMPW is a mixed bed scouting column for water-soluble linear polymers. The pore volume can be accessed by polymers ranging in molecular weight from 500 to 8,000,000 Da. By quickly categorizing the MW profile of an unknown sample, the column enables a fast selection of the best TSKgel PW column for routine analysis. Mixed bed columns are also used when a sample has a very broad MW distribution.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service