About This Item
Recommended Products
mp
94-96 °C (lit.)
storage temp.
2-8°C
SMILES string
CN1C(=O)C=CC1=O
InChI
1S/C5H5NO2/c1-6-4(7)2-3-5(6)8/h2-3H,1H3
InChI key
SEEYREPSKCQBBF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service