Skip to Content
Merck
All Photos(2)

Key Documents

HPA059010

Sigma-Aldrich

Anti-CACNG3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
PLN 2,680.00

PLN 2,680.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
PLN 2,680.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

PLN 2,680.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PSKITMGTLLNSDRDHAFLQFHNSTPKEFKESLHNNPAN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNG3(10368)

General description

Calcium voltage-gated channel auxiliary subunit γ 3 (CACNG3) is a human neuronal voltage-gated calcium channel. The protein belongs to calcium channel γ subunit family. It is expressed in brain and retina. The gene is located on human chromosome 16p12-p13.1.

Immunogen

calcium channel, voltage-dependent, gamma subunit 3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calcium voltage-gated channel auxiliary subunit γ 3 (CACNG3) regulates α-amino-3-hydroxyl-5-methyl-4-isoxazole-propionic acid (AMPA) receptor localization. The protein is associated with childhood absence epilepsy (CAE). Mutations in CACNG3 is associated with calcium channel dysfunction, nervous hereditary disease and retinal disease. Calcium channels such as CACNG3 and CACNG2 maintains calcium homeostasis, regulates Ca2+ dependant actions such as muscular contraction, releasing of hormone and neurotransmitter and signal transduction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86258

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linkage and association analysis of CACNG3 in childhood absence epilepsy
Everett KV, et al.
European Journal of Human Genetics, 15(4), 463-463 (2007)
Identification and characterization of human neuronal voltage-gated calcium channel gamma 3 subunit gene
Xia J, et al.
Chinese Science Bulletin, 45(23), 2172-2176 (2000)
D L Burgess et al.
Genome research, 9(12), 1204-1213 (1999-12-30)
Gene duplication is believed to be an important evolutionary mechanism for generating functional diversity within genomes. The accumulated products of ancient duplication events can be readily observed among the genes encoding voltage-dependent Ca(2+) ion channels. Ten paralogous genes have been
Ren-Shiang Chen et al.
Cell biochemistry and biophysics, 47(2), 178-186 (2007-07-27)
The calcium channel gamma subunits comprise an eight-member protein family that share a common topology consisting of four transmembrane domains and intracellular N- and C-termini. Although the first gamma subunit was identified as an auxiliary subunit of a voltage-dependent calcium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service