Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C19H14N2O2
CAS Number:
Molecular Weight:
302.33
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
assay
≥95% (HPLC)
form
solid
solubility
DMSO: 12 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
CC(=O)c1ccc(Nc2c3ccccc3nc4occc24)cc1
InChI
1S/C19H14N2O2/c1-12(22)13-6-8-14(9-7-13)20-18-15-4-2-3-5-17(15)21-19-16(18)10-11-23-19/h2-11H,1H3,(H,20,21)
InChI key
VJDPGTFIMDGXDQ-UHFFFAOYSA-N
Biochem/physiol Actions
CIL-102 is a tubulin polymerization inhibitor: apoptosis inducer.
Legal Information
US Patent No. 6,750,223. Sold for non-human research purposes under a non-exclusive license agreement with Kaohsiung Medical University.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥95% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| form solid | form solid | form powder | form powder |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 12 mg/mL | solubility DMSO: 30 mg/mL, water, high purity: 30 mg/mL | solubility DMSO: 10 mg/mL, clear | solubility DMSO: 30 mg/mL, clear |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wen-Shih Huang et al.
PloS one, 12(1), e0168989-e0168989 (2017-01-10)
CIL-102 (1-[4-(furo[2,3-b]quinolin-4-ylamino)phenyl]ethanone) is a well-known, major active agent of the alkaloid derivative of Camptotheca acuminata with valuable biological properties, including anti-tumorigenic activity. In this study, we investigated the molecular mechanisms by which CIL-102 mediated the induction of cell death, and
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C5492-10MG | 04061833502228 |



