Skip to Content
Merck
All Photos(1)

Key Documents

BM0034

Sigma-Aldrich

BMS-764459

≥98% (HPLC)

Synonym(s):

(S)-4-(1-Cyclopropyl-2-methoxyethyl)-6-(6-(difluoromethoxy)-2,5-dimethylpyridin-3-ylamino)-5-oxo-4,5-dihydropyrazine-2-carbonitrile, 4-[(1S)-1-Cyclopropyl-2-methoxyethyl]-6-[[6-(difluoromethoxy)-2,5-dimethyl-3-pyridinyl]amino]-4,5-dihydro-5-oxo-2-pyrazinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21F2N5O3
CAS Number:
Molecular Weight:
405.40
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C1C(NC2=CC(C)=C(OC(F)F)N=C2C)=NC(C#N)=CN1[C@H](COC)C3CC3

InChI

1S/C19H21F2N5O3/c1-10-6-14(11(2)23-17(10)29-19(20)21)25-16-18(27)26(8-13(7-22)24-16)15(9-28-3)12-4-5-12/h6,8,12,15,19H,4-5,9H2,1-3H3,(H,24,25)/t15-/m1/s1

InChI key

PJMUNXORSUNUCV-OAHLLOKOSA-N

Biochem/physiol Actions

BMS-764459 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1) with high affinity (IC50 = 0.86 nM against 150 pM ovine CRF for binding rat CRF-R1), potency (IC50 = 1.9 nM against 1 nM CRF-stimulated cAMP production in human Y-79 retinoblastoma cells), and selectivity, displaying little affinity toward CRF-R2/CRF2 and 43 other receptor/channel/transporter proteins (IC50 >10 μM). BMS-764459 exhibits anxiolytic efficacy in a rat defensive withdrawal model of anxiety in vivo (1-3 mg/kg p.o.) with good oral bioavailability (F in dogs = 53% with 2 mg/kg in suspension and 70% with 3 mg/kg in solution).
BMS-764459 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1).

Features and Benefits

BMS-764459 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damir Simic et al.
Toxicology, 311(3), 169-177 (2013-07-09)
The corticotrophin releasing factor (CRF) receptor I antagonist, BMS-764459 (evaluated as a potential treatment of affective disorders), was orally dosed to female Sprague-Dawley rats once daily for 2 weeks (vehicle control or 175mg/kg/day). To investigate the mechanism of BMS-764459-related liver
Richard A Hartz et al.
Journal of medicinal chemistry, 52(23), 7653-7668 (2009-12-04)
Detailed metabolic characterization of 8, an earlier lead pyrazinone-based corticotropin-releasing factor-1 (CRF(1)) receptor antagonist, revealed that this compound formed significant levels of reactive metabolites, as measured by in vivo and in vitro biotransformation studies. This was of particular concern due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service