Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C12H21N5O4S
CAS Number:
Molecular Weight:
331.39
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
assay
>95% (HPLC)
form
solid
storage temp.
−20°C
Biochem/physiol Actions
Aldehyde reactive probe used to selectively modify abasic sites of DNA.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.