About This Item
description
non-ionic
Quality Level
assay
≥99.0% (TLC)
form
solid
optical activity
[α]20/D +52.5±3°, c = 0.5% in methanol
mol wt
452.49 g/mol
technique(s)
protein purification: suitable, protein quantification: suitable
CMC
120 mM (20-25°C), 120
SMILES string
OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OCCC3CCCCC3)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C20H36O11/c21-8-11-13(23)14(24)16(26)20(29-11)31-18-12(9-22)30-19(17(27)15(18)25)28-7-6-10-4-2-1-3-5-10/h10-27H,1-9H2/t11-,12-,13-,14+,15-,16-,17-,18-,19-,20-/m1/s1
InChI key
RHXPDNGQJSXOMW-OIIXUNCGSA-N
Application
Legal Information
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
A guide displaying detergents' specifications and applications. The detergents are seperated into four groups (nonionic, anionic, cationic, zwitterionic) based on their hydrophillic head group. The guide also contains Non-Detergent Sulfobetaines.