Skip to Content
Merck

Skip To

12203C

Fetal Bovine Serum

New Zealand origin, sterile-filtered, suitable for cell culture

Synonym(s):

FBS, FCS, sera, serum

Sign In to View Organizational & Contract Pricing.

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
Biological source:
bovine fetus
Origin:
New Zealand origin
Sterility:
sterile-filtered
Shipped in:
dry ice


biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Premier

composition

hemoglobin, ≤25 mg/dL

origin

New Zealand origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Application

FBS sourced from New Zealand is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Other Notes

(Formerly product number F4922)


Still not finding the right product?

Explore all of our products under Fetal Bovine Serum


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



David B Finlay et al.
PeerJ, 4, e1835-e1835 (2016-03-29)
The orphan receptor GPR18 has become a research target following the discovery of a putative endogenous agonist, N-arachidonoyl glycine (NAGly). Chemical similarity between NAGly and the endocannabinoid anandamide suggested the hypothesis that GPR18 is a third cannabinoid receptor. GPR18-mediated cellular
Robert C Tuckey et al.
The Journal of steroid biochemistry and molecular biology, 112(4-5), 213-219 (2008-11-13)
Cytochrome P450scc (CYP11A1) metabolizes vitamin D3 to 20-hydroxyvitamin D3 as the major product, with subsequent production of dihydroxy and trihydroxy derivatives. The aim of this study was to determine whether cytochrome P450scc could metabolize 1alpha-hydroxyvitamin D3 and whether products were
Cynthia K Hahn et al.
Cancer cell, 16(4), 281-294 (2009-10-06)
Cell-based screening can facilitate the rapid identification of compounds inducing complex cellular phenotypes. Advancing a compound toward the clinic, however, generally requires the identification of precise mechanisms of action. We previously found that epidermal growth factor receptor (EGFR) inhibitors induce



Global Trade Item Number

SKUGTIN
12203C-500ML04061838720313
12203C-1000ML04061838243904
12203C-50ML04061838243911
12203C-100ML04061838719867

Questions

1–2 of 2 Questions  
  1. Once a bottle of Fetal Bovine Serum (FBS) is thawed, for how long can the bottle be used?

    1 answer
    1. Once Fetal Bovine Serum has been thawed, the bottle can be used in cell culture for up to 7 days for most cell types when stored  refrigerated at 2-8°C.  If the cells are not fastidous (e.g. HeLa or fibroblasts), the thawed and refrigerated serum can be used for up to 30 days.  For some extremely fastidious cell lines, it may be necessary to screen individual lots for acceptable performance.  For uses such as blocking for various immuno procedures, usage will generally be for 30 days and perhaps somewhat longer when stored at 2-8°C.  For other procedures, usage beyond 30 days storage at 2-8°C will need to be determined.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters