Skip to Content
Merck

Skip To

114-500

Endothelial Cell Serum-Free, Growth Factor-Free Medium (500 ml)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
PLN 651.00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

PLN 651.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

A proprietary medium developed to induce morphological differentiation of CADMEC in Attachment Factor Solution coated tissue culture ware in 50% confluent culture. This medium induces CADMEC to form cord-like structures in 48 hours. These differentiated cells will resume their cobblestone morphology when switched back to CADMEC Growth Medium. It is supplemented with defined components to allow angiogenesis studies on the Attachment Factor-coated tissue culture ware without the interference of undefined components in Matrigel.

Preparation Note

contained in protocol

Analysis Note

Each lot was tested for the ability to support plating, spreading and proliferation

Legal Information

CADMEC is a trademark of Cell Applications

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
114PR-500113-500113PR-500
form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shelf life

6 mo.

shelf life

6 mo.

shelf life

6 mo.

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
114-50004061838701602
MTCB5780-500EA04065272291606
MTCB5790-500EA04065272282352
MTCB5771-500EA04065272282338

Questions

Reviews

No rating value

Active Filters