Skip to Content
Merck

Skip To

MGVS16

Medical Viscosity Standard

viscosity 1.6 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
PLN 322.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

PLN 322.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

1.6 mPa.s(25.00 °C)

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

storage temp.

2-8°C

General description

Find detailed technical data for all Paragon Medical Viscosity Standards on the Paragon Scientific webpage
These products contain antibacterial agent.
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 under UKAS accreditation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MGVS12MGVS20MGVS40
format

mixture

format

mixture

format

mixture

format

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

viscosity

1.6 mPa.s(25.00 °C)

viscosity

1.2 mPa.s(25.00 °C)

viscosity

2.0 mPa.s(25.00 °C)

viscosity

4.0 mPa.s(25.00 °C)

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?

Explore all of our products under Medical Viscosity Standard


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MGVS16-500ML04061837680724
MGVS16-100ML04061837680717

Questions

Reviews

No rating value

Active Filters