Skip to Content
Merck

Skip To

ST1703

Anti-DLL1 Mouse mAb (4F8)

liquid, clone 4F8, Calbiochem®

Synonym(s):

Anti-Delta-like 1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Clone:
4F8, monoclonal
Species reactivity:
human
Application:
Citations:
-


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F8, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLL1(28514)

General description

Anti-DLL1, mouse monoclonal, clone 4F8, recognizes the DLL1 protein in HeLa cells. It is validated for use in ELISA, Western blotting, and immunocytochemistry.
Purified mouse monoclonal antibody. Recognizes the DLL1 protein.
Recognizes the DLL1 protein in HeLa cells.

Immunogen

A recombinant polypeptide corresponding to amino acids of 18-110 human DLL1, expressed as a GST fusion protein

Application



ELISA (see coments)
Immunoblotting (1-5 g/ml)
Immunocytochemistry (10 g/ml)

Physical form

In PBS, pH 7.2.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HeLa cells

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 &micro/g/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters