Skip to Content
Merck

Skip To

BA24

Anti-Thrombospondin (Ab-3) Mouse mAb (A6.1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Available to ship TODAYfromKuehne + Nagel Sp. z o.o.

PLN 1,930.00

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(bovine, canine, horse, human, porcine, rat, sheep)
Species reactivity:
-
Application:
Citations:
-

PLN 1,930.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

form

liquid

species reactivity

(bovine, canine, horse, human, porcine, rat, sheep)

storage condition

OK to freeze

dilution

(Electron Microscopy
Neutralization Studies
Flow Cytometry
Frozen Sections (1:50-1:100)
Immunoblotting (1:200-1:400)
Immunofluorescence
Paraffin Sections (not recommended))

isotype

IgG1

storage temp.

−20°C

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/Ag14 mouse myeloma cells. Recognizes the ~450 kDa (non-reduced) or the ~180 kDa (reduced) thrombospondin (TSP) protein.

Immunogen

Human

Application

Electron Microscopy Neutralization Studies Flow Cytometry Frozen Sections (1:50-1:100)Immunoblotting (1:200-1:400)Immunofluorescence Paraffin Sections (not recommended)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
BA24-100UG04055977220735