Skip to Content
Merck

Skip To

AC112

Sheep anti-Human IgG Antibody

1 mg/mL, from sheep

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
1.050,00 PLN

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
ELISA
Citations:
-

1.050,00 PLN


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins which stage immune responses against pathogenic organisms. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fcg receptors which interact with sialylated Fc fragments on IgG molecules.
~150 kDa observed.

Immunogen

Purified human IgG

Application

ELISA Analysis: As little as 0.025 µg/mL of this antibody detected Human IgG (IgG>IgM>IgG Fc>IgG Fab>IgA>IgM Fc).
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
Sheep anti-Human IgG Antibody detects level of Sheep Human IgG & has been published & validated for use in Sheep Human IgG.

Biochem/physiol Actions

This antibody demonstrates minimal to no cross reactivity against Mouse IgG, Rabbit IgG, Rat IgG, Hamster IgG, Guinea Pig IgG, and Chicken IgY.
This antibody demonstrates specificity with Human IgG (IgG>IgM>IgG Fc>IgG Fab>IgA>IgM Fc).

Physical form

Format: Semi-Purified
Octanoic Acid and Ammonium Sulfate cut then depleted
Purified sheep polyclonal antibody in buffer containing PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by ELISA in a panel of antibodies containing Mouse IgG, Rabbit IgG, Rat IgG, Hamster IgG, Guinea Pig IgG, and Human IgG.

ELISA Analysis: As little as 0.025 µg/mL of this antibody detected Human IgG with minimal to no cross-reactivty against Mouse IgG, Rabbit IgG, Rat IgG, Hamster IgG and Guinea Pig IgG.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AP512AC111BAC112C
description

1 mg/mL, from sheep

description

Chemicon®, from sheep

description

from sheep, biotin conjugate

description

from sheep, CY3 conjugate

biological source

sheep

biological source

sheep

biological source

sheep

biological source

sheep

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

immunohistochemistry: suitable

technique(s)

immunocytochemistry: suitable

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

species reactivity

human

species reactivity

human

species reactivity

mouse

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters