Skip to Content
Merck

Skip To

69871

pET-29a(+) DNA - Novagen

Novagen′s pET-29a-c(+) vectors carry an N-terminal S•Tag /thrombin configuration plus an optional C-terminal His•Tag sequence.

Synonym(s):

cloning vector, expression vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 μg

Estimated to ship onApril 01, 2026fromKuehne + Nagel Sp. z o.o.

PLN 1,600.00

About This Item

UNSPSC Code:
41106600
NACRES:
NA.51

PLN 1,600.00


Estimated to ship onApril 01, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
699526974069741
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Storage Class

10 - Combustible liquids

wgk

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
69871-304055977258752

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service