Merck
69749
pET-24a(+) DNA - Novagen
Novagen′s pET-24a-d(+) vectors carry an N-terminal T7-Tag sequence plus an optional C-terminal His-Tag sequence.
Synonym(s):
cloning vector, expression vector, vector
Sign In to View Organizational & Contract Pricing.
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41106610
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Visit the Document Library
Global Trade Item Number
SKUGTIN
C6645-1G04061826744178
C6645-25MG04061833511442
C6645-500MG04061833511459
C6645-100MG04061833511435
C6645-5G04061833511466
69749-250G04061835570652
69749-50G04061832787367
69749-1KG04061832787336