Skip to Content
Merck
All Photos(1)

Key Documents

APO455836496

Apollo Scientific Ltd

2-Fluoro-5-methoxyphenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
165.17
MDL number:
Pricing and availability is not currently available.

Iupac Name

(2-fluoro-5-methoxyphenyl)acetonitrile

Inchi Code

1S/C9H8FNO/c1-12-8-2-3-9(10)7(6-8)4-5-11/h2-3,6H,4H2,1H3

InChI key

QSPYWCDXOCAMGL-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxyphenylacetonitrile

Synonym

2-Fluoro-5-methoxybenzyl cyanide, (2-Fluoro-5-methoxyphenyl)acetonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxyphenylacetonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library