Skip to Content
Merck
All Photos(1)

Key Documents

160741

Sigma-Aldrich

3-Methyl-2-nitrophenol

99%

Synonym(s):

2-Nitro-m-cresol, 3-Hydroxy-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
2047479
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

106-108 °C/9.5 mmHg (lit.)

mp

35-39 °C (lit.)

functional group

nitro

SMILES string

Cc1cccc(O)c1[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-3-2-4-6(9)7(5)8(10)11/h2-4,9H,1H3

InChI key

QIORDSKCCHRSSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-2-nitrophenol is also referred as 3-methyl-2-nitro-1-hydroxybenzene.[1] FT-IR and FT-Raman spectra of 3-methyl-2-nitrophenol has been studied.[2]

Application

3-Methyl-2-nitrophenol was used to investigate the formation of nitrous acid (HONO) in the gas phase in a flow tube photoreactor upon irradiation of ortho-nitrophenols.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iustinian Bejan et al.
Physical chemistry chemical physics : PCCP, 8(17), 2028-2035 (2006-04-25)
Formation of nitrous acid (HONO) in the gas phase has been observed for the first time in a flow tube photoreactor upon irradiation (lambda = 300-500 nm) of 2-nitrophenol and methyl substituted derivatives using a selective and sensitive instrument (LOPAP)
D Mahadevan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 139-151 (2011-11-15)
In the present study, the FT-IR and FT-Raman spectra of 3-methyl-2-nitrophenol (C(7)H(7)O(3)N) (3M2NP) have been recorded in the range of 4000-100 cm(-1). The fundamental modes of vibrational frequencies of 3M2NP are assigned. All the geometrical parameters have been calculated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service