Skip to Content
Merck
All Photos(1)

Key Documents

40100-U

Supelco

α-BHC solution

certified reference material, 1000 μg/mL in methanol

Synonym(s):

α-HCH solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H

InChI key

JLYXXMFPNIAWKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pernilla Carlsson et al.
Journal of environmental monitoring : JEM, 14(5), 1437-1443 (2012-04-07)
A pilot study is presented evaluating selected chlorinated pesticides as chemical tracers for water masses in a sub-Arctic fjord system (Godthåbsfjord, western Greenland). Polyoxymethylene (POM) based passive water samplers were deployed during summer-autumn 2010. The levels of the analysed chlorinated
Sung-Deuk Choi et al.
Environmental science & technology, 45(20), 8834-8841 (2011-09-13)
An understanding of the factors that control the time trends of persistent organic pollutants (POPs) in the environment is required to evaluate the effectiveness of emission reductions and to predict future exposure. Using a regional contaminant fate model, CoZMo-POP 2
M Pućko et al.
Environmental science & technology, 46(21), 11862-11869 (2012-10-09)
During the summer of 2009, we sampled 14 partially refrozen melt ponds and the top 1 m of old ice in the pond vicinity for α-hexachlorocyclohexane (α-HCH) concentrations and enantiomer fractions (EFs) in the Beaufort Sea. α-HCH concentrations were 3
Esha Sharma et al.
European journal of obstetrics, gynecology, and reproductive biology, 161(2), 163-169 (2012-02-09)
Organochlorine pesticides (OCPs) and oxidative stress are reported to be associated with adverse reproductive outcomes. Glutathione S-transferase (GST) is a polymorphic supergene family involved in the detoxification of numerous toxins including OCPs. OCPs are endocrine disrupter and prenatal exposure to
Gaoxin Zhou et al.
Chirality, 24(8), 615-620 (2012-06-08)
The enantioselective bioaccumulation and elimination behaviors of α-hexachlorocyclohexane (α-HCH) enantiomers in earthworm and soil were investigated by chiral gas chromatography. Enantiomer fraction values were calculated as indicators of the enantioselectivity. The mature earthworms were exposed to 0.10 µg g(-1)(wwt) (0.14

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service