Skip to Content
Merck
α-BHC solution certified reference material, 1000 μg/mL in methanol

α-BHC solution

α-BHC solution certified reference material, 1000 μg/mL in methanol
Synonym(s):
α-HCH solution
Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83

Products

Product Category
Brand
Formula Weight
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing