Skip to Content
Merck
All Photos(1)

Key Documents

W524905

Sigma-Aldrich

Dipentene

mixture of isomers

Synonym(s):

(±)-Limonene, p-Mentha-1,8-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
3587825
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.001
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

vapor density

>4.7 (80 °C, vs air)

vapor pressure

1 mmHg ( 20 °C)

autoignition temp.

458 °F

expl. lim.

0.7-6.1 %, 150 °F

refractive index

n20/D 1.473 (lit.)

bp

170-180 °C (lit.)

density

0.86 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

citrus

SMILES string

CC(=C)C1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3

InChI key

XMGQYMWWDOXHJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad M Rafiee Fanood et al.
Nature communications, 6, 7511-7511 (2015-06-25)
Simultaneous, enantiomer-specific identification of chiral molecules in multi-component mixtures is extremely challenging. Many established techniques for single-component analysis fail to provide selectivity in multi-component mixtures and lack sensitivity for dilute samples. Here we show how enantiomers may be differentiated by
K Vanommeslaeghe et al.
Journal of chemical information and modeling, 52(12), 3144-3154 (2012-11-14)
Molecular mechanics force fields are widely used in computer-aided drug design for the study of drug-like molecules alone or interacting with biological systems. In simulations involving biological macromolecules, the biological part is typically represented by a specialized biomolecular force field
Keng-Chih Liu et al.
International journal of pharmaceutics, 441(1-2), 687-692 (2012-10-27)
Gap junction protein connexin43 (Cx43) specific antisense oligodeoxynucleotides (AsODN) have been shown to improve a number of inflammatory conditions and may therefore offer a novel strategy for persistent pain management. However, for such molecules to be clinically effective, delivery challenges
Patrizia A d'Alessio et al.
Life sciences, 92(24-26), 1151-1156 (2013-05-15)
To further explore the anti-inflammatory properties of d-Limonene. A rat model was used to compare evolution of TNBS (2,5,6-trinitrobenzene sulfonic acid)-induced colitis after oral feeding with d-Limonene compared to ibuprofen. Peripheral levels of TNF-α (Tumor Necrosis Factor alpha) were assessed
Young Woo Kim et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 16(1), 17-38 (2013-04-12)
d-Limonene, a major constituent of citrus oils, is a monoterpene widely used as a flavor/fragrance additive in cosmetics, foods, and industrial solvents as it possesses a pleasant lemon-like odor. d-Limonene has been designated as a chemical with low toxicity based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service