Skip to Content
Merck
Dipentene mixture of isomers

Dipentene

Dipentene mixture of isomers
Synonym(s):
(±)-Limonene, p-Mentha-1,8-diene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing