Skip to Content
Merck
All Photos(2)

Documents

148245

Sigma-Aldrich

2-Methoxynaphthalene

99%

Synonym(s):

Methyl 2-naphthyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
Beilstein:
1859408
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

274 °C (lit.)

mp

70-73 °C (lit.)

solubility

benzene: soluble
carbon disulfide: soluble
diethyl ether: soluble

density

1.064 g/mL at 25 °C (lit.)

SMILES string

COc1ccc2ccccc2c1

InChI

1S/C11H10O/c1-12-11-7-6-9-4-2-3-5-10(9)8-11/h2-8H,1H3

InChI key

LUZDYPLAQQGJEA-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

2-methoxynaphthalene acylation was used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria L Blair et al.
Inorganic chemistry, 48(18), 8863-8870 (2009-09-15)
Alkali-metal-mediated manganation (AMMMn) reactions of the synergic base sodium monoalkyl-bisamidomanganate [(tmeda)Na(tmp)(CH(2)SiMe(3))Mn(tmp)] (1) with naphthalene, 1-methoxynaphthalene, or 2-methoxynaphthalene are reported. These novel direct manganation [Mn(II)] reactions produced the crystalline ortho-manganated naphthyl products [(tmeda)Na(tmp)(2-C(10)H(7))Mn(tmp)] (3), [(tmeda)Na(tmp){2-(1-MeOC(10)H(6))}Mn(CH(2)SiMe(3))] (4), and [(tmeda)Na(tmp){3-(2-MeOC(10)H(6))}Mn(tmp)] (5) in reasonable
Xiaoying Ouyang et al.
Journal of the American Chemical Society, 136(4), 1449-1461 (2013-12-19)
Layered borosilicate zeolite precursor ERB-1P (Si/B = 11) is delaminated via isomorphous substitution of Al for B using a simple aqueous Al(NO3)3 treatment. Characterization by PXRD shows loss of long-range order, and TEM demonstrates transformation of rectilinear layers in the
P Di Gennaro et al.
Biotechnology and bioengineering, 93(3), 511-518 (2005-09-30)
The bioconversion of naphthalene to the 1,2-dihydro-1,2-dihydroxy derivative was performed in good yield using an Escherichia coli recombinant strain carrying Pseudomonas fluorescens N3 dioxygenase. However, the efficiency of such transformation is affected by many process parameters, and their optimization is
G M Whited et al.
Bioorganic & medicinal chemistry, 2(7), 727-734 (1994-07-01)
2-Methoxynaphthalene was subjected to biooxidation by whole cells of six organisms: Pseudomonas putida F39/D containing toluene dioxygenase, Escherichia coli JM109(pDTG601), containing recombinant toluene dioxygenase from Pp F39/D, Pseudomonas sp. NCIB 9816/11, containing naphthalene dioxygenase. E. coli JM109(pDTG141), containing recombinant naphthalene
H Widén et al.
Food additives and contaminants, 22(7), 681-692 (2005-07-16)
Mineral water and soft drinks with a perceptible off-odour were analysed to identify contaminants originating from previous misuse of the refillable polyethylene terephthalate (PET) bottle. Consumers detected the off-odour after opening the bottle and duly returned it with the remaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service