Skip to Content
Merck
2-Methoxynaphthalene 99%

2-Methoxynaphthalene

2-Methoxynaphthalene 99%
Synonym(s):
Methyl 2-naphthyl ether
Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing