Permanent gases and light hydrocarbons
Materiales
columna analítica
column
Carboxen-1006 PLOT, 30 m × 0.53 mm I.D. (25461)
column
Carboxen 1004 micropacked, 2 m × 1/16 in. stainless steel (12846)
oven
35 °C (1 min), 24 °C/min to 225 °C (micropacked)
oven
35 °C (5 min), 24 °C/min to 225 °C (PLOT)
detector
TCD, 230 °C
carrier gas
helium, 22 cm/sec
injection
35 μL
sample
1% each analyte in nitrogen
Descripción
Información legal
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany