Merck
Expression of ATP-Binding Cassette Transporters B1 and C1 after Severe Traumatic Brain Injury in Humans.
Journal of neurotrauma (2015-04-22)
F Anthony Willyerd, Philip E Empey, Ashley Philbrick, Milos D Ikonomovic, Ava M Puccio, Patrick M Kochanek, David O Okonkwo, Robert S B Clark
RESUMEN
Adenosine triphosphate-binding cassette (ABC) transport proteins ABCC1 and ABCB1 (also known as multidrug resistance-associated protein 1 and p-glycoprotein, respectively), are key membrane efflux transporters of drugs and endogenous substrates, including in the brain. The impact of traumatic brain injury (TBI) on ABCC1 and ABCB1 expression in humans is unknown. We hypothesized that ABCC1 and ABCB1 expression would be altered in brain tissue from patients acutely after severe TBI. Archived TBI samples (n=10) from our Brain Trauma Research Center and control samples (n=7) from our Alzheimer Disease Research Center were obtained under Institutional Review Board approval. Protein was extracted from fresh frozen cortical brain tissue for Western blot analysis and sections were obtained from fixed cortical tissue for immunohistochemistry. Relative abundance of ABCC1 was increased in samples from TBI versus controls (2.8±2.5 fold; p=0.005). ABCC1 immunohistochemistry was consistent with Western blot data, with increased immunoreactivity in cerebral blood vessel walls, as well as cells with the morphological appearance of neurons and glia in TBI versus controls. Relative abundance of ABCB1 was similar between TBI and controls (p=0.76), and ABCB1 immunoreactivity was primarily associated with cerebral blood vessels in both groups. These human data show that TBI increases ABCC1 expression in the brain, consistent with possible implications for both patients receiving pharmacological inhibitors and/or substrates of ABCC1 after TBI.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cóctel de inhibidores de proteasas, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti--actina antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC1