Documentos clave
EP0030119827
Eppendorf® Tube Rack for 5.0 and 15 mL Tubes
Holds 12 x 5-15 mL tubes, 12 wells (2 rows of 6 wells each), pkg of 2 pieces, white, autoclavable
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
material
polypropylene
feature
autoclavable
12 wells (2 rows of 6 wells each)
Holds 12 x 5-15 mL tubes
packaging
pkg of 2 pieces
manufacturer/tradename
Eppendorf® 30119827
color
white
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)