Skip to Content
Merck
All Photos(1)

Documents

SML1577

Sigma-Aldrich

Rubrofusarin

≥98% (HPLC)

Synonym(s):

5,6-Dihydroxy-8-methoxy-2-methyl-benzo[g]chromen-4-one, CID 72537, NSC258316

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow to orange

storage temp.

−20°C

InChI

1S/C15H12O5/c1-7-3-10(16)14-12(20-7)5-8-4-9(19-2)6-11(17)13(8)15(14)18/h3-6,17-18H,1-2H3

InChI key

FPNKCZKRICBAKG-UHFFFAOYSA-N

Biochem/physiol Actions

Rubrofusarin is an orange polyketide pigment produced by Fusarium graminearum and other fungal species that exhibit potent anti-cancer and anti-mycobacterial activities. Rubrofusarin is a mycotoxin that inhibits human DNA topoisomerase II-α.. Additionally, Rubrofusarin shows a significant anti-estrogenic activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service