SMB01033
Bis(methylthio)gliotoxin (FR-49175)
≥90% (LC/MS-ELSD)
Synonym(s):
Bisdethiobis(methylthio)gliotoxin, S,S-Dimethyl gliotoxin, dimethylgliotoxin
About This Item
Recommended Products
biological source
natural
Assay
≥90% (LC/MS-ELSD)
form
solid
mol wt
356.46
solubility
water: soluble
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
InChI
1S/C15H20N2O4S2/c1-16-12(20)14(22-2)7-9-5-4-6-10(19)11(9)17(14)13(21)15(16,8-18)23-3/h4-6,10-11,18-19H,7-8H2,1-3H3/t10-,11-,14+,15+/m0/s1
InChI key
OVBAGMZLGLXSBN-UOVKNHIHSA-N
Related Categories
General description
Application
Features and Benefits
- Suitable for Biochemical and Biomedical research
- Versatile and adaptable for wide variety of laboratory and research applications
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service