Recommended Products
form
liquid
Quality Level
packaging
pkg of 500 mL
manufacturer/tradename
Montage®
technique(s)
molecular cloning: suitable (BAC prep)
molecular cloning: suitable (plasmid miniprep)
shipped in
ambient
General description
The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) minipreps
Application
Research Category
All
All
Suitable for neutralization in plasmid & BAC minipreps
Storage and Stability
Reagents should be stored at 15 °C to 30 °C.
Legal Information
MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service