Skip to Content
Merck
All Photos(1)

Key Documents

29578-U

DIN6 Gas Cylinder Pressure Regulator

2-stage high purity plus, parameter 0-150 psi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

parameter

0-150 psi

Looking for similar products? Visit Product Comparison Guide

General description

For use with helium, nitrogen, or argon gas cylinders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this regulator rated for use with hydrogen? Do you have a cut sheet, or additional information to provide?

    1 answer
    1. This product is used with helium, nitrogen, or argon gas cylinders. It is not tested for suitability with hydrogen.

      Regulator types:
      DIN6 - For use with helium, nitrogen, or argon gas cylinders
      DIN1 - For use with hydrogen, methane, or 5% methane in argon gas cylinders

      Please see the link: https://www.genstartech.com/sites/default/files/Specialty%20Gas%20Regulators%20%26%20Accessories.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service