Skip to Content
Merck
All Photos(1)

Key Documents

SRP6414

Sigma-Aldrich

Cathepsin B human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

APPS, CPSB, CTSB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 29 kDa (Mature)
calculated mol wt 36.7 kDa (Pro)
observed mol wt 34 kDa
observed mol wt 43_kDaand34 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTSB(1508)

General description

Cathepsin B (CTSB) also known as APP secretase (APPS) and CPSB, is an enzymatic protein belonging to the peptidase C1 family. Cathepsin B / CTSB is synthesized as a preproenzyme. Following removal of the signal peptide, the inactive proenzyme undergoes further modifications including removal of the pro region to result in the active enzyme. The catalytic activity of Cathepsin B / APPS contains: Hydrolysis of proteins with broad specificity for peptide bonds; preferentially cleaves -Arg-Arg-|-Xaa bonds in small molecule substrates (thus differing from cathepsin L); In addition to being an endopeptidase, shows peptidyl-dipeptidase activity, liberating C-terminal dipeptides. As a thiol protease, cathepsin B / CPSB is believed to participate in intracellular degradation and turnover of proteins and has also been implicated in tumor invasion and metastasis. Overexpression of cathepsin B has been associated with esophageal adenocarcinoma and other tumors.
DTT-reduced. Protein migrates due to glycosylation. Arg18 (pro) or Phe74 (mature) is the predicted N-terminal.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM Tris-HCl and 150 mM NaCl, pH 8.0. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service