Skip to Content
Merck
All Photos(1)

Key Documents

21263

Supelco

GC Stationary Phase

phase SP®-1200, bottle of 10 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

packaging

bottle of 10 g

parameter

25-200 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

SP®-1200 phase

solubility

chloroform: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients.[1] The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered in order to choose a stationary phase.[2] SP-1200 is a low polarity ester type developed to separate the C2-C5 free acids in aqueous solution.[3]
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

SP 1200 is suitable in GC analysis for quantitative analysis of volatile fatty acids.[4]

Legal Information

SP is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanley Blackburn
Peptides, 286-286 (1986)
A E van den Bogaard et al.
Journal of clinical microbiology, 23(3), 523-530 (1986-03-01)
Gas chromatographic analysis of volatile fatty acids for identification of obligately anaerobic bacteria and for presumptive diagnosis of anaerobic infections is now widely practiced. However, it is difficult to compare data because only a qualitative analysis is done or only
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service