Key Documents
Z692697
microDOC Compact gel documentation system, with UV Transilluminator (UVT254)
AC/DC input 110 V AC
Select a Size
1 EA
€5,830.00
€5,830.00
Estimated to ship on14 March 2025Details
Select a Size
Change View
1 EA
€5,830.00
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
€5,830.00
Estimated to ship on14 March 2025Details
manufacturer/tradename
Cleaver CSL-MDOCUV254$
General description
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254 nm tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254 nm transilluminator
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)