Key Documents
VIV105007STU3
SynVivo SynTumour 3D cancer model chip, microvascular network configuration, 2 um pillar
new
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
Pricing and availability is not currently available.
packaging
case of 3 ea
manufacturer/tradename
SynVivo 105007-STU3
storage temp.
room temp
General description
SynTumor cancer model chip- Microvascular design Barrier type: Pillars, Distance between pillars: 3 μm, Pillar Diameter: 20 μm, Barrier height: 2 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.
Linkage
LInk to Publications application text
Choose from one of the most recent versions:
Certificates of Analysis (COA)