Skip to Content
Merck
All Photos(1)

Key Documents

V5757

Sigma-Aldrich

Vernolic acid methyl ester

≥99%

Synonym(s):

(+)-(12S,13R)-Epoxy-cis-9-octadecenoic acid methyl ester, Linoleic acid 12:13-oxide methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H34O3
CAS Number:
Molecular Weight:
310.47
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99%

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@@H]1O[C@@H]1C\C=C/CCCCCCCC(=O)OC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service