Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702340

Sigma-Aldrich

Anti-Thyroglobulin Antibody, clone 3I10H1, Rabbit Monoclonal

Synonym(s):

AITD3, TGN

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€379.00

€379.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€379.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€379.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

3I10H1, monoclonal

form

liquid

species reactivity

human, rat, rat

concentration

0.70 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200

color

colorless

isotype

IgG

immunogen sequence

STLSFYQRRRFSPDDSAGASALLRSGPYMPQCDAFGSWEPVQCHAGTGHCWCVDEKGGFIPGSLTARSLQIPQCPTTCEKSRTSGLLSSWKQARSQENPSP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TG(7038)

General description

Thyroglobulin (Tg) is a glycoprotein homodimer produced predominantly by the thryroid gland. It acts as a substrate for the synthesis of thyroxine and triiodothyronine as well as the storage of the inactive forms of thyroid hormone and iodine. Thyroglobulin is secreted from the endoplasmic reticulum to its site of iodination, and subsequent thyroxine biosynthesis, in the follicular lumen. Mutations in this gene cause thyroid dyshormonogenesis, manifested as goiter, and are associated with moderate to severe congenital hypothyroidism. Polymorphisms in this gene are associated with susceptibility to autoimmune thyroid diseases (AITD) such as Graves disease and Hashimoto thryoiditis. [provided by RefSeq, Nov 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human Thyroglobulin (P01266).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

IHC, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service