Skip to Content
Merck
All Photos(3)

Key Documents

SAB1407766

Sigma-Aldrich

Anti-DDX24 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90.3 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX24(57062)

Related Categories

General description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which shows little similarity to any of the other known human DEAD box proteins, but shows a high similarity to mouse Ddx24 at the amino acid level. (provided by RefSeq)

Immunogen

DDX24 (AAH96826.1, 1 a.a. ~ 804 a.a) full-length human protein.

Sequence
MKLKDTKSRPKQSSCGKFQTKGIKVVGKWKEVKIDPNMFADGQMDDLVCFEELTDYQLVSPAKNPSSLFSKEAPKRKAQAVSEEEEEEEGKSSSPKKKIKLKKSKNVATEGTSTQKEFEVKDPELEAQGDDMVCDDPEAGEMTSENLVQTAPKKKKNKGKKGLEPSQSTAAIRDKLDILGAAETGSGKTLAFAIPMIHAVLQWQKRNAAPPPSNTEAPPGETRTEAGAETRSPGKAEAESDALPDDTVIESEALPSDIAAEARAKTGGTVSDQALLFGDDDAGEGPSSLIREKPVPKQNENEEENLDKEQTGNLKQELDDKSATCKAYPKRPLLGLVLTPTRELAVQVKQHIDAVARFTGIKTAILVGGMSTQKQQRMLNRRPEIVVATPGRLWELIKEKHYHLRNLRQLRCLVVDEADRMVEKGHFAELSQLLEMLNDSQYNPKRQTLVFSATLTLVHQAPARILHKKHTKKMDKTAKLDLLMQKIGMRGKPKVIDLTRNEATVETLTETKIHCETDEKDFYLYYFLMQYPGRSLVFANSISCIKRLSGLLKVLDIMPLTLHACMHQKQRLRNLEQFARLEDCVLLATDVAARGLDIPKVQHVIHYQVPRTSEIYVHRSGRTARATNEGLSLMLIGPEDVINFKKIYKTLKKDEDIPLFPVQTKYMDVVKERIRLARQIEKSEYRNFQACLHNSWIEQAAAALEIELEEDMYKGGKADQQEERRRQKQMKVLKKELRHLLSQPLFTESQKTKYPTQSGKPPLLVSAPSKSESALSCLSKQKKKKTKKPKEPQPEQPQPSTSAN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service