Skip to Content
Merck
All Photos(1)

Key Documents

PZ0355

Sigma-Aldrich

PF-05085727

≥98% (HPLC)

Synonym(s):

4-(1-Azetidinyl)-1-methyl-3-[1-methyl-5-[4-(trifluoromethyl)phenyl]-1H-pyrazol-4-yl]-1H-pyrazolo[3,4-d]pyrimidine, 4-(Azetidin-1-yl)-1-methyl-3-(1-methyl-5-(4-(trifluoromethyl)phenyl)-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-d]pyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€81.40
25 MG
€295.00

€81.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€81.40
25 MG
€295.00

About This Item

Empirical Formula (Hill Notation):
C20H18F3N7
CAS Number:
Molecular Weight:
413.40
UNSPSC Code:
51111800
NACRES:
NA.77

€81.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CN(N=C1)C(C2=CC=C(C(F)(F)F)C=C2)=C1C3=NN(C)C4=C3C(N5CCC5)=NC=N4

InChI key

SHAVEYUXRBIMLA-UHFFFAOYSA-N

Biochem/physiol Actions

PF-05085727 is a potent inhibitor against cGMP-dependent phosphodiesterase PDE2 (PDE2A; IC50 = 2.3 nM) with >500-fold selectivity over other PDEs. Its 3-[(18)F]fluoroazetidinyl derivative ([(18)F]PF-05270430) has been successfully applied via i.v. injection as a brain-permeant probe for imaging brain PDE2 in live cynomolgus monkeys by positron emission tomography (PET).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhang et al.
Journal of medicinal chemistry, 56(11), 4568-4579 (2013-05-09)
To accelerate the discovery of novel small molecule central nervous system (CNS) positron emission tomography (PET) ligands, we aimed to define a property space that would facilitate ligand design and prioritization, thereby providing a higher probability of success for novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service