M3678
EX-CELL® MDCK Production Medium
Animal-component free, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Synonym(s):
Madin-Darby Canine Kidney Protein-free Medium
Select a Size
€775.00
Select a Size
About This Item
€775.00
description
for research or for further manufacturing use
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
storage temp.
2-8°C
Application
Components
Suitability
Reconstitution
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service