Skip to Content
Merck
All Photos(1)

Key Documents

D4822

Sigma-Aldrich

Dulbecco′s Modified Eagle′s Medium - high glucose

HEPES modification, With 4500 mg/L glucose, L-glutamine, 25 mM HEPES, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, DMEM

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352207

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

impurities

Endotoxin, tested

components

phenol red: yes
L-glutamine: yes
sodium pyruvate: no
glucose: high

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frances Gays et al.
PloS one, 6(3), e18475-e18475 (2011-04-13)
Comprehensive analysis of the transcription start sites of the Ly49 genes of C57BL/6 mice using the oligo-capping 5'-RACE technique revealed that the genes encoding the "missing self" inhibitory receptors, Ly49A, C, G, and I, were transcribed from multiple broad regions
Elena Seiradake et al.
The EMBO journal, 30(21), 4479-4488 (2011-09-29)
Brain wiring depends on cells making highly localized and selective connections through surface protein-protein interactions, including those between NetrinGs and NetrinG ligands (NGLs). The NetrinGs are members of the structurally uncharacterized netrin family. We present a comprehensive crystallographic analysis comprising
Haiqiang Wu et al.
PloS one, 9(10), e109438-e109438 (2014-10-03)
Inhibition of β-amyloid (Aβ) aggregation is an attractive therapeutic and preventive strategy for the discovery of disease-modifying agents in Alzheimer's disease (AD). Phomopsis occulta is a new, salt-tolerant fungus isolated from mangrove Pongamia pinnata (L.) Pierre. We report here the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service