Skip to Content
Merck
All Photos(1)

Key Documents

MABC527

Sigma-Aldrich

Anti-PLGF Antibody, clone 3A6.2 | MABC527

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

antibody form

purified antibody

Quality Level

clone

3A6.2
monoclonal

purified by

using protein G

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG2b

UniProt accession no.

General description

GST-tagged recombinant protein corresponding to human PLGF.

Application

Epitope: Near C-terminus

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected PLGF in 10 µg of HUVEC cell lysate.

Physical properties

~25 kDa observed

Physical form

Purified mouse monoclonal IgG2bλ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service